ContactUsLinkComponent
Klinger Beer Lambert's Law Experiment Complete Set Up
Klinger Beer Lambert's Law Experiment Complete Set Up
Catalog # 470356-832
Supplier:  Klinger
New Product
undefined
Klinger Beer Lambert's Law Experiment Complete Set Up
Catalog # 470356-832
Supplier:  Klinger
Supplier Number:  KSCIBLL
New Product
Specifications
  • Dimensions:
    Beer-Lambert's law apparatus
  • Cat. no.:
    470356-832
Specifications
About this item
Find the concentration of a liquid using the samples of known concentration using the Beer-Lambert law.
  • Precise optical alignment: The optical alignment of the components is attained by optical bench, the setting up time is faster and experimentation is easy
  • Digital Lux meter: This enables measurement of light intensity at various angles of reflection. The meter reads with a precision of 1 lux
The Beer Lambert law relates the attenuation of light through a substance and the properties of that substance. Light interacts with matter in the following ways: emission, absorption, transmission, and reflection or scattering. Depending on the physical and chemical properties of the matter under interaction, there can be one or more ways in which light interacts. It is because of these interactions light can be used as a probe to measure the physical and chemical properties of materials.
Ordering information: Kit includes (1) optical bench set 0.8 m, (2) uprights, (1) light source holder, (1) polarizer holder, (1) analyzer holder, (1) adjustable collimating slit holder, (1) cuvette holder, (1) light sensor holder, (1) data processor, (1) power supply for light source, (1) glass cuvette.