ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Catalog # 77461-568
Supplier: AAT BIOQUEST INC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Size:1 ml
- Environmentally Preferable:
- Cat. no.:77461-568
Specifications
About this item
10 mM dNTP Mix is a mixture of four nucleotides (dATP, dCTP, dGTP, dTTP) in purified water.
- Each nucleotide is at a concentration of 10 mM
- The formulation is stable for 12 months at −20 °C
10 mM dNTP mix is suitable for use in polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.
Caution: For research use only.