ContactUsLinkComponent
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-530
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 488-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-530
Supplier:  AAT BIOQUEST INC
Supplier Number:  17044
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    100 nmoles
  • Cat. no.:
    77461-530
Specifications
About this item
This iFluor 488-dUTP conjugate is used as a green fluorescence color with SpectrumGreen™ filter set (SpectrumGreen™ is the trademark of Vysis).
  • Widely used for both FISH probes and microarray-based experiments
  • Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
Caution: For research use only.