Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7.5)*
Catalog # 77461-518
Supplier: AAT BIOQUEST INC
CAS Number:
Specifications
- Size:25 nmoles
- Cat. no.:77461-518
Specifications
About this item
Fluorescein-12-dCTP might be incorporated into a DNA sequence as its natural counterpart dCTP to incorporate a fluorescein tag in a DNA molecule.
- Can be used for the direct enzymatic labeling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and cytidine has been optimized for the optimal substrate incorporation and labeling efficiency.
Caution: For research use only.