iFluor® 488 PEG12-dUTP *1 mM in TE Buffer (pH 7.5)*
Catalog # 77461-524
Supplier: AAT BIOQUEST INC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:25 nmoles
- Cat. no.:77461-524
Specifications
About this item
This iFluor® 488-PEG12-dUTP conjugate has the same spectral properties to ChromaTide™ Alexa Fluor 488-dUTP.
- PEG12 spacer makes it much more effectively incorporated into DNAs by polymerase
- Bright signal that has high photostability and is not affected by pH
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via the conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labeling or PCR.
Caution: For research use only.