ContactUsLinkComponent
Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7.5)*
Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7.5)*
Catalog # 77461-520
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7.5)*
Catalog # 77461-520
Supplier:  AAT BIOQUEST INC
Supplier Number:  17036
CAS Number:  
Specifications
  • Size:
    25 nmoles
  • Cat. no.:
    77461-520
Specifications
About this item
Fluorescein-12-dATP might be incorporated into a DNA sequence as its natural counterpart dATP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labeling of DNA/cDNA, e.g., by PCR or Nick Translation
The linker between fluorescein tag and adenine has been optimized for the optimal substrate incorporation and labeling efficiency.
Caution: For research use only.