Orders LinkContactUsLinkComponent
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
Supplier:   AAT Bioquest
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
Supplier:  AAT Bioquest
Certificates
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
ReadiUse™ Annexin Binding Buffer, Optimized for Imaging and Flow Cytometry
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The ReadiUse™ Annexin Binding Buffer *Optimized for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
  • Provided as a 10X concentrate
  • Saves time
  • pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
 : For research use only.
Product Family Options
Product Information
  • Availability
    Price
  • 77461-918
    ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 10 ml
    ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 10 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $137.18
    $137.18
    Each(1/Itm/Bx)
       $137.18
    Supplier Number: 22870
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 77461-920
    ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 ml
    ReadiUse™ Annexin binding buffer, optimized for imaging and flow cytometry, 100 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $173.57
    $173.57
    Each(1/Itm/Bx)
       $173.57
    Supplier Number: 22871
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.