HPLC Supply and Waste Justrite Bundle
Supplier: SCAT AMERICAS, INC.
New Product
Certificates
About this item
HPLC Supply and waste starter bundle, includes HPLC starter kit 3 V2.0, suitable for all HPLC systems.
Certifications: UN approved.
Delivery: Kit includes 4×SafetyCap III, 8×blind plug, 12×fitting 1,6/2,3/3,2 mm OD, 4×air valve, 4 Laboratory bottle DURAN, GL 45, 1000 ml, clear glass and round, with cap and pouring ring, Exhaust filter M, V3.0, with splash guard and change indicator, service life 6 months, HPLC Manifold with stainless steel and PP Quick Disconnect for Justrite containers, PTFE-EL and PTFE, electrostatic conductive, 6×1,6/2,3/3,2 mm AD, 6×PFA-blindplug, 6×5,0 to 11,5 mm ID, 6×PTFE-blindplug, Adapter for exhaust filter to stainless steel coupling and CPC coupling, for Justrite container.
Frequently Bought Together