Order Entry
Canada
ContactUsLinkComponent
 
Human TrkC Luciferase Reporter Assay Kit
 
Human TrkC Luciferase Reporter Assay Kit

 

  • Storage conditions:
    Do not store in liquid nitrogen. Reporter cells cannot be refrozen or maintained in extended culture.
  • Species reactivity:
    Human
  • Storage temperature:
    −80 °C

 

Frequently Bought Together