Order Entry
Canada
ContactUsLinkComponent
Human ADRB1 Luciferase Reporter Assay Kit
Human ADRB1 Luciferase Reporter Assay Kit
Certificates
Human ADRB1 Luciferase Reporter Assay Kit

Specifications

  • Storage conditions:
    Do not store in liquid nitrogen. Reporter cells cannot be refrozen or maintained in extended culture.
  • Species reactivity:
    Human
  • Storage temperature:
    −80 °C

Specifications

Frequently Bought Together