- Description:Adrenoreceptor alpha 1A (ADRA1A) reporter assay kit
- Storage conditions:Do not store in liquid nitrogen. Reporter cells cannot be refrozen or maintained in extended culture.
- Species reactivity:Human
- Storage temperature:−80 °C
- Cat. no.:104087-942
- No. of Wells:384
- No. of Assays/Kit:1
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO's adrenoreceptor alpha 1A (ADRA1A) reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against ADRA1A.
- Includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor
- Kit includes reporter cells, optimized media for use during cell thawing and in diluting test samples, reference agonist, luciferase detection reagent, cell culture-ready assay plate, and detailed protocol
- Exceptional cell viability post-thaw
- Consistent results lot to lot (product quality data available upon request)
This is an all-inclusive cell-based luciferase reporter assay kit targeting the human adrenoreceptor alpha 1A (ADRA1A). INDIGO's ADRA1A reporter assay utilizes proprietary mammalian cells that have been engineered to provide constitutive expression of the ADRA1A. In addition to ADRA1A reporter cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, a reference agonist, luciferase detection reagent, and a cell culture-ready assay plate. The principal application of this assay is in the screening of test samples to quantify any functional activity, either agonist or antagonist, that they may exert against ADRA1A. This kit provides researchers with: Clear, reproducible results, exceptional cell viability post-thaw. Consistent results lot to lot.