AccuThrive® DermaBlade® Biopsy Blade
77587-128
New Product
- Description:DermaBlade® biopsy blade, sterile
- Material:Stainless steel
- Thickness:0.004"
- Cat. no.:77587-128
AccuThrive® DermaBlade® is a one-piece shave biopsy instrument designed by a dermatologist for cutaneous surgery.
- MicroCoat® stainless steel blade
- Sure grip sides provide stability and control
- Enhanced user safety with no-touch, ready to use blade
- Flexible to remove either surface protuberances or deep lesions
- Sterile, individually packaged
- Disposable, single-use device
DermaBlade® for shave biopsy is designed to provide the flexibility to remove surface protuberances or deep lesions while maintaining control throughout the incision. DermaBlade® also features MicroCoat® Enrichment Technology which utilizes a proprietary polymer coating to enhance precision and reduce friction, providing a smooth resistance-free excision.
: Blades are extremely sharp. Do not use if package is damaged. Bending blades may break them. Handle with care. Used sharps are contaminated. Disease may occur from contact or injury. Dispose in puncture-resistant container. Refer to local guidelines for handling and disposal of sharps.