Order Entry
Canada
ContactUsLinkComponent
Kinase Dilution Buffers VII
Kinase Dilution Buffers VII
Catalog # 77558-950
Supplier:  Sino Biological
New Product
CAS Number:  
Kinase Dilution Buffers VII
Catalog # 77558-950
Supplier:  Sino Biological
Supplier Number:  K27-09-05
New Product
CAS Number:  

Specifications

  • Description:
    Kinase dilution buffer VII
  • Size:
    5 ml
  • Storage temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 5 mM MOPS, pH 7.2, 2.5 mM B-glycerol-phosphate, 5 mM MgCl₂, 1 mM EGTA, 0.4 mM EDTA, 0.05 mM DTT, 50 ng/μl BSA, 5% glycerol
  • Cat. no.:
    77558-950

Specifications

About this item

Solution of buffer used for diluting kinase protein.

For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.