Methyltransferase Assay Buffer IV
Supplier: Sino Biological
New Product
Certificates
About this item
Stock solution of the buffer used for assaying protein methyltransferases.
Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.
Specifications
- Storage temperature:–20 °C
- Formulation:Frozen solution of buffer containing: 100 mM Tris-HCl, pH 8.0, 250 mM NaCl, 5 mM EDTA, 15 mM MgCl₂, 0.5 mg/ml BSA.
Specifications
Frequently Bought Together