Order Entry
Canada
ContactUsLinkComponent
Methyltransferase Assay Buffer III
Methyltransferase Assay Buffer III
Supplier:  Sino Biological
New Product
Certificates
Methyltransferase Assay Buffer III
Supplier:  Sino Biological
New Product

Specifications

  • Storage temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 7.5, 5 mM EDTA, 25% Glycerol, 50 ng/µl BSA.

Specifications

Related Information

Frequently Bought Together