Order Entry
Canada
ContactUsLinkComponent
Methyltransferase Assay Buffer II
Methyltransferase Assay Buffer II
Supplier:  Sino Biological
New Product
Certificates
Methyltransferase Assay Buffer II
Supplier:  Sino Biological
New Product

Specifications

  • Storage temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 8.0 and 50 ng/µl BSA

Specifications

Related Information

Frequently Bought Together