Order Entry
Canada
Orders LinkContactUsLinkComponent
Methyltransferase Assay Buffer I

Methyltransferase Assay Buffer I

Supplier:   Sino Biological
Methyltransferase Assay Buffer I
Supplier:  Sino Biological
Certificates

3 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Stock solution of the buffer used for assaying protein methyltransferases.

Storage: For optimal storage, aliquot diluted product into smaller quantities and store at recommended temperature. For most favorable performance, avoid repeated handling and multiple freeze/thaw cycles.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77559-336
  • 77559-338
  • 77559-340
  • Formulation:
  • Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
  • Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
  • Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
  • Storage temperature:
  • –20 °C
  • –20 °C
  • –20 °C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 77559-336
    Buffer for enzyme assay
    Buffer for enzyme assay
    1 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $27.49
    $27.49
    Each(1/Itm/Bx)
       $27.49
    Minimum order quantity is 1

    Supplier Number: M01-09-01
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77559-336
    Formulation
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
    Storage temperature
    –20 °C
  • 77559-338
    Buffer for enzyme assay
    Buffer for enzyme assay
    5 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $81.66
    $81.66
    Each(1/Itm/Bx)
       $81.66
    Minimum order quantity is 1

    Supplier Number: M01-09-05
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77559-338
    Formulation
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
    Storage temperature
    –20 °C
  • 77559-340
    Buffer for enzyme assay
    Buffer for enzyme assay
    20 ml
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $171.04
    $171.04
    Each(1/Itm/Bx)
       $171.04
    Minimum order quantity is 1

    Supplier Number: M01-09-20
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77559-340
    Formulation
    Frozen solution of buffer containing: 250 mM Tris-HCl, pH 9.0 and 50 ng/µl BSA.
    Storage temperature
    –20 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.