Orders LinkContactUsLinkComponent
Logic® Telescoping Base Stands
Logic® Telescoping Base Stands
Supplier:   Labconco
Logic® Telescoping Base Stands
Supplier:  Labconco
Certificates
Logic® Telescoping Base Stands
Logic® Telescoping Base Stands
8 Options Available Below
Product Details & Documents
Telescoping base stand for Labconco Class II biosafety cabinets, fully welded design, optic white with toe-locking casters and fixed leveling feet.
  • Fully Welded design
  • For optic white Logic BSC models
  • Supports loads up to 4,000 lbs.
  • Castered models supports loads up to 1,000 lbs.
  • Durable 1.75" tubular, epoxy-coated steel frame
  • Telescoping legs are adjustable to eight height positions at installation in 2.5 cm (1") increments to provide a working height from 76 cm (30") to 94 cm (37")
  • Available with either fixed leveling feet or 5" easy-clean casters with toe locks
Certifications: NSF approved.
Product Family Options
With fixed feet
Product Information
  • Dimensions
    Availability
    Price
  • 77574-556
    3' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    3' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    97.8W×73.7D×69.9 - 87.6H cm (38.5× 29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $1,730.00
    $1,730.00
    Each
       $1,730.00
    Supplier Number: 5173300
  • 77574-566
    4' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    4' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    128.3W×73.7D×69.9 - 87.6H cm (50.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $1,820.00
    $1,820.00
    Each
       $1,820.00
    Supplier Number: 5173400
  • 77574-576
    5' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    5' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    158.8W×73.7D×69.9 - 87.6H cm (62.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $1,850.00
    $1,850.00
    Each
       $1,850.00
    Supplier Number: 5173500
  • 77574-586
    6' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    6' Telescoping base stand, optic white, fully welded design with fixed leveling feet
    189.2W×73.7D×69.9 - 87.6H cm (74.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $1,920.00
    $1,920.00
    Each
       $1,920.00
    Supplier Number: 5173600
With toe-locking casters
Product Information
  • Dimensions
    Availability
    Price
  • 77574-558
    3' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    3' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    97.8W×73.7D×69.9 - 87.6H cm (38.5× 29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $2,150.00
    $2,150.00
    Each
       $2,150.00
    Supplier Number: 5173310
  • 77574-568
    4' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    4' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    128.3W×73.7D×69.9 - 87.6H cm (50.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $2,250.00
    $2,250.00
    Each
       $2,250.00
    Supplier Number: 5173410
  • 77574-578
    5' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    5' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    158.8W×73.7D×69.9 - 87.6H cm (62.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $2,290.00
    $2,290.00
    Each
       $2,290.00
    Supplier Number: 5173510
  • 77574-588
    6' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    6' Telescoping base stand, optic white, fully welded design with with toe-locking casters
    189.2W×73.7D×69.9 - 87.6H cm (74.5×29×27.5 - 34.5")
    Log in for availability
    Log in for availability
    Each
    Each - $2,350.00
    $2,350.00
    Each
       $2,350.00
    Supplier Number: 5173610