HPLC Supply and Waste Starter Bundle and Bundle PLUS, Type 20
Catalog # 77553-648
Supplier: SCAT AMERICAS, INC.
New Product
Specifications
- Capacity:20 L
- Description:HPLC Supply and waste starter bundle, Thread size: S60/61
- Material:HDPE
- Cat. no.:77553-648
Specifications
About this item
HPLC Supply and waste starter bundle with labelling field, V2.0, suitable for all HPLC systems.
Certifications: UN approved.
Delivery: Supplied with 1× Safety cap III starter kit, V2.0, GL 45, 1×safety waste cap, V2.0, S60/61, 1×exhaust filter M, V3.0, with splash guard and change indicator, 1×20 L canister, S60/61, PE-HD.