Chicken Interferon Alpha (IFNa) ELISA Kit
Catalog # 77536-900
Supplier: Bioss
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Chicken
- Target protein:Interferon Alpha (IFNa)
- Size:1 kit
- Detection method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf life:12 Months
- Detection range:78.1 - 5000 pg/ml
- Storage temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:28 pg/ml
- Cat. no.:77536-900
- No. of Tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results