Human C-Raf Proto Oncogene Serine/Threonine Protein Kinase (CRAF) ELISA Kit
Catalog # 77538-134
Supplier: Bioss
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:C-Raf Proto Oncogene Serine/Threonine Protein Kinase (CRAF)
- Size:1 kit
- Detection method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf life:12 Months
- Detection range:0.156 - 10 ng/ml
- Storage temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:0.055 ng/ml
- Cat. no.:77538-134
- No. of Tests:48
Specifications
About this item
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results