Bovine Anti-Mullerian Hormone (AMH) ELISA Kit
77536-620
: Bioss
New Product
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Primary antibody reactivity:Bovine
- Target protein:Anti-Mullerian Hormone (AMH)
- Size:1 kit
- Detection method:Colorimetric microplate reader primary wavelength 450 nm
- Shelf life:12 Months
- Detection range:0.312 - 20 ng/ml
- Storage temperature:TMB substrate, wash buffer (30X concentrate), and the stop solution should be stored at 4 °C upon receipt, while the other items should be stored at –20 °C.
- Sensitivity:0.117 ng/ml
- Cat. no.:77536-620
- No. of Tests:48
- High sensitivity with a broad assay range
- Flexible inputs: Serum, plasma, cell culture supernatant
- Reproducible and specific results