Order Entry
Canada
Orders LinkContactUsLinkComponent
Human IL-5 ELISA Kit

Human IL-5 ELISA Kit

2 units
Catalog # MSPP-02010
Marketsource
Supplier #:  02010
Human IL-5 ELISA Kit
Catalog # MSPP-02010
Supplier:  
Marketsource
CAS Number:  
Human IL-5 ELISA Kit
Human IL-5 ELISA Kit

2 units

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For detection and measurement of human interleukin 5.

The Human Interleukin 5 (IL-5) ELISA Kit is designed for the quantitative detection and measurement of human IL-5 in biological fluids such as serum, plasma, and cell culture supernatants. IL-5 is produced by Th2 cells, eosinophils, and activated mast cells. IL-5 plays an important role in the recruitment, activation, proliferation, and survival of eosinophils. The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine.

The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample. The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Size:
    2 units
  • Cat. No.:
    MSPP-02010
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-5

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.