Order Entry
Canada
ContactUsLinkComponent
Human IFN-gamma ELISA Kit
Human IFN-gamma ELISA Kit
Catalog # MSPP-02002
CAS Number:  
undefined
Human IFN-gamma ELISA Kit
Catalog # MSPP-02002
Supplier Number:  02002
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IFN-gamma
  • Size:
    2 units
  • Cat. no.:
    MSPP-02002

Specifications

About this item

For detection and measurement of human interferon-gamma.

The Human Interferon-gamma (IFN-γ) ELISA Kit is designed for the quantitative detection and measurement of human IFN-γ in biological fluids such as serum, plasma, and cell culture supernatants. IFN-γ is primarily secreted by T and NK cells, but is also produced by dendritic cells and macrophages in smaller amounts. IFN-γ is a pleiotropic cytokine that modulates a broad range of immunological functions including antimicrobial and antitumor activity, and increases the surface expression of MHC class I and II proteins.

The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine. The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample. The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').