Order Entry
Canada
ContactUsLinkComponent
 
Methyltransferase Dilution Buffers II
New Product
 
Methyltransferase Dilution Buffers II
New Product

 

  • Storage temperature:
    Store product at –20 °C
  • Formulation:
    Frozen solution of buffer containing: 50 mM Tris-HCl, pH 8.0 and 10 ng/µl BSA

 

Frequently Bought Together