Brady® Self-Laminating Cryogenic Polyester Labels
About this item
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides.
- Permanent acrylic adhesive
- Matte finish
- Autoclavable, optically clear material is suitable for cryogenic storage
- Recommended ribbon: R4300 (CA94000-380)
Dependable material is able to withstand extreme temperatures from −196 to +130 °C (−320 to +266 °F), immersion in liquid nitrogen, and autoclave use. Labels are resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% clorox solution. Made with a clear overlaminate that protects the print and maintains content visibility.
3 Options Available Below
Product Details & Documents
Versatile labels are ideal for curved surfaces such as vials, centrifuge tubes, test tubes, straws and slides.
- Permanent acrylic adhesive
- Matte finish
- Autoclavable, optically clear material is suitable for cryogenic storage
- Recommended ribbon: R4300 (CA94000-380)
Dependable material is able to withstand extreme temperatures from −196 to +130 °C (−320 to +266 °F), immersion in liquid nitrogen, and autoclave use. Labels are resistant to ethanol, toluene, isopropanol, xylene, DMSO, 50% acetic acid, 10% hydrochloric acid, 10% clorox solution. Made with a clear overlaminate that protects the print and maintains content visibility.
Product Family Options
Product Information
- MaterialColorFor Use WithDimensionsPackagingAvailabilityPrice
- PolyesterBlack on white/clearBMP41;BMP51;BMP53;M5110.6H×1.8"W180 labels/rollLog in for availabilityEach$115.25Supplier Number: 170769
- Supplier Number: 170765
- PolyesterBlack on white/clearBMP51;BMP53;M5111H × 1.75"W180 labels/rollLog in for availabilityEach$115.25Supplier Number: 170878