Order Entry
Canada
ContactUsLinkComponent
 
Kinase Assay Buffers II
New Product
 
Kinase Assay Buffers II
New Product

 

  • Form:
    Liquid
  • Storage temperature:
    –20 °C
  • Formulation:
    Frozen solution of buffer containing: 25 mM MOPS, pH 7.2, 12.5 mM B-glycerol-phosphate, 20 mM MgCl₂, 12.5 mM MnCl₂, 5 mM EGTA, 2 mM EDTA, 0.25 mM DTT.

 

Frequently Bought Together