Supertek® Variable Inertia Bar
Catalog # 470356-596
Supplier: Shiv Dial Sud & Sons
New Product
Specifications
- Description:Variable inertia bar
- Environmentally Preferable:
- Cat. no.:470356-596
Specifications
About this item
A classic demonstration of the dependence of angular acceleration on moment of inertia.
- Support stand and weights available separately
This apparatus helps to demonstrate that the torque required to accelerate a flywheel depends upon its moment of inertia rather than its mass. A steel bar (790×12 mm) is mounted at its center on a sleeved spindle. The spindle is extended through a sleeve so that the bar may be rotated by a weight and string. Heavy adjustable masses are mounted on each end of the bar and maybe clamped at any desired position, enabling inertia to be varied.