About this item
The SHOWA 383 is the world’s-first biodegradable multi-purpose glove! Its reduced thickness ensures the 383 optimum tactility when working on delicate projects. In addition, the design provides comfort and flexibility during extended wear. The 383 is manufactured with the Eco Best Technology®, making it biodegradable.
- General purpose
- Thin palm material
- Thin finger thickness
- Embossed microporous nitrile coating
- Lightweight and flexible
- Engineered yarn
- Reusable and Biodegradable
The gloves are abrasion and oil resistant, latex free with increased dexterity and sensitivity. It has an optimal liner design.
Trades and Applications: Intricate parts handling, light assembly of oil-coated pieces, mechanical and engineering, automotive repairs and maintenance, shuttering, component assembly, heavy equipment operation, tool operation and transporting.
Industries: Construction, mechanical, aerospace, automotive, engineering, mining, airports and ports.
Specifications
- Catalog No:
- 77575-156
- 77575-108
- 77575-116
- 77575-114
- 77575-142
- Model:
- 383
- 383
- 383
- 383
- 383
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Material:
- Yarn
- Yarn
- Yarn
- Yarn
- Yarn
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded