ContactUsLinkComponent
Vacuum Seal for Rotavapor® Rotary Evaporators, VS22
  77569-848
 :  Buchi
New Product
undefined
Vacuum Seal for Rotavapor® Rotary Evaporators, VS22
  77569-848
 :  Buchi
 :  11075810
New Product
  • Description:
    VS22 vacuum seal
  • Cat. no.:
    77569-848
  • For Use With:
    Rotavapor® R-80, R-100, R-1×4
Ensure efficient operation of your Rotavapor® by replacing the vacuum seal as needed. Designed specifically for BUCHI Rotavapor® and glassware with high chemical resistance.
  • High chemical resistance
  • OEM seal for proper fit and performance
Vacuum seal VS22 PTFE base, NBR O-ring, FDA-compliant. Compatible with R-80, R-100, R-1×4.