Orders LinkContactUsLinkComponent
SteriWare® Scoops, Long Handled
New Product
SteriWare® Scoops, Long Handled
New Product
Certificates
SteriWare® Scoops, Long Handled
SteriWare® Scoops, Long Handled
7 Options Available Below
Product Details & Documents
These SteriWare® long handled scoops are the ideal tool for reaching into deep containers. The extra long handle will help operators keep their hands out of the product.
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Available sterile (SAL 10⁻⁶) and non sterile
They are all manufactured and packed in a cleanroom to ensure cleanliness and reduce particulate contamination.
These single use scoops are ideal for use in the laboratory, production and warehouse.
Handle length: 300 mm.
 : Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials. BSE-/TSE-free. Full batch traceability.
 : Individually bagged.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Width
    Sterility
    Availability
    Price
  • HDPE
    100 ml
    390 mm
    50 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $236.10
    $236.10
    Case of 50
       $236.10
    Supplier Number: 8341H-100
    Specifications:
    New Product
    New Product
  • HDPE
    100 ml
    390 mm
    50 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $339.29
    $339.29
    Case of 50
       $339.29
    Supplier Number: 8341H-100S
    Specifications:
    New Product
    New Product
  • HDPE
    25 ml
    356 mm
    34 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $236.10
    $236.10
    Case of 50
       $236.10
    Supplier Number: 8341H-25
    Specifications:
    New Product
    New Product
  • HDPE
    25 ml
    356 mm
    34 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $339.29
    $339.29
    Case of 50
       $339.29
    Supplier Number: 8341H-25S
    Specifications:
    New Product
    New Product
  • HDPE
    50 ml
    371 mm
    40 mm
    Non sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $236.10
    $236.10
    Case of 50
       $236.10
    Supplier Number: 8341H-50
    Specifications:
    New Product
    New Product
  • HDPE
    50 ml
    371 mm
    40 mm
    Sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $339.28
    $339.28
    Case of 50
       $339.28
    Supplier Number: 8341H-50S
    Specifications:
    New Product
    New Product
  • 100 ml
    Sterile
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $365.39
    $365.39
    Case of 50
       $365.39
    Supplier Number: 8341R-100S
    Specifications:
    New Product
    New Product