Orders LinkContactUsLinkComponent
CRISPRclean® Stranded Total RNA Prep
CRISPRclean® Stranded Total RNA Prep
CRISPRclean® stranded total RNA prep, 24 reactions
Catalog # CA77575-232
Supplier #:  NOVA-512830
CRISPRclean® Stranded Total RNA Prep
Catalog # CA77575-232
CAS Number:  
CRISPRclean® Stranded Total RNA Prep
CRISPRclean® Stranded Total RNA Prep
CRISPRclean® stranded total RNA prep, 24 reactions
CAS Number:  
Price
6 133,44 $
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The CRISPRclean® stranded total RNA prep with rRNA depletion (HMR) is a total RNA library prep leveraging a CRISPR-powered workflow facilitating the detection of lower expressing and biologically relevant transcripts from complex samples.
  • Efficient multi-species depletion using CRISPR-Cas9 technology in a single tube
  • Enhances detection of biologically relevant, low-expressing transcripts
  • Optimized library prep and depletion workflow ensures high-quality transcript representation
  • Achieve consistent full-length, uniform transcript coverage with high library complexity
  • Complete workflows in just one day: 7.5 hours assay with 3 hours of hands-on time
Specific, unbiased rRNA depletion and library prep for RNA-Seq. In RNA samples, up to 90% consists of ribosomal RNA. Failing to remove these abundant rRNA sequences during transcriptome sequencing reduces sequencing capacity and hinders the detection of biologically relevant, low-expressing transcripts. Discover how CRISPRclean® enhances research by efficiently removing rRNA.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Description:
    CRISPRclean® stranded total RNA prep
  • Size:
    24 reactions