Order Entry
Canada
ContactUsLinkComponent
Anti-KLRG1 Hamster Monoclonal Antibody [clone: 2F1] (APC)
Anti-KLRG1 Hamster Monoclonal Antibody [clone: 2F1] (APC)
Catalog # MSPP-100-1632
undefined
Anti-KLRG1 Hamster Monoclonal Antibody [clone: 2F1] (APC)
Catalog # MSPP-100-1632
Supplier Number:  100-1632

Specifications

  • Antigen name:
    KLRG1
  • Clonality:
    Monoclonal
  • Clone:
    2F1
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Hamster
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Size:
    100 µg
  • Gene ID:
    50928
  • Cat. no.:
    MSPP-100-1632

Specifications

About this item

Hamster (Syrian) monoclonal antibody against mouse KLRG1, APC-conjugated.

  • Nk cells
  • T cells CD8

This monoclonal antibody (clone 2F1) reacts with the mouse killer cell lectin-like receptor G1 (KLRG1), a 30 - 38 kDa homodimer receptor and a member of the lectin-like type 2 transmembrane receptor family of proteins. This receptor is expressed by activated, mature natural killer (NK) cells and by effector and memory T cells. KLRG1 expression has been associated with reduced proliferative capacity of activated T lymphocytes or reduced effector functions of activated NK cells. It has also been noted that cell surface expression of KLRG1 is upregulated by expression of the major histocompatibility (MHC) class I molecules, and this can be mediated by interactions with class I-specific Ly49 inhibitory receptors. The expression of mouse KLRG1, using 2F1 antibody clone, has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. Use this recombinant monoclonal antibody in applications requiring detection of KLRG1, such as flow cytometry (FC).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: APC
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse