To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antigen name:TNF-alpha
- Clonality:Monoclonal
- Clone:MT15B15
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 Tests
- Environmentally Preferable:
- Gene ID:7124
- Cat. no.:MSPP-100-1461
Mouse monoclonal antibody against human TNF-alpha, PF647P-conjugated.
- Macrophages
- T cells
This mouse monoclonal antibody (clone MT15B15) reacts with human tumor necrosis factor alpha (TNF-α). TNF-α is a pro-inflammatory cytokine primarily produced by macrophages and T cells in response to infection and inflammation. Initially generated as a ~26 kDa transmembrane precursor, TNF-α is then processed by metalloproteinases into a ~17 kDa soluble product. Both forms of TNF-α are active in trimeric form and exert their effects via binding to either TNFR1, present on the surface of nearly all human cell types, or TNFR2, which is localized to immune and endothelial cells. First described as having tumor-necrotic action, TNF-α signaling results in an inflammatory response that includes vasodilation and edema formation, adhesion of leukocytes to the epithelium, and blood coagulation. TNF-α also plays a key role in defense against bacterial, viral, and parasitic infections.
Type:
Antigen: TNF-alpha
Clonality: Monoclonal
Clone: MT15B15
Conjugation: PF647P
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human