Order Entry
Canada
ContactUsLinkComponent
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2]
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2]
Catalogue # MSPP-60036
Fournisseur:  STEMCELL Technologies
undefined
Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2]
Catalogue # MSPP-60036
Fournisseur:  STEMCELL Technologies
Supplier Number:  60036

Spécifications

  • Antigen name:
    CD150 (SLAM)
  • Clonality:
    Monoclonal
  • Clone:
    TC15-12F12.2
  • Host:
    Rat
  • Isotype:
    IgG2a lambda
  • Reactivity:
    Mouse
  • Size:
    500 µg
  • Gene ID:
    27218
  • Cat. no.:
    MSPP-60036

Spécifications

À propos de cet article

Rat monoclonal IgG2a antibody against mouse CD150 (SLAM), unconjugated.

  • Hematopoietic stem
  • Progenitor cells

The TC15-12F12.2 antibody reacts with CD150 (signaling lymphocyte activation molecule or SLAM), an ~75 kDa type I transmembrane glycoprotein that plays multiple roles in the immune response by serving as a cell adhesion molecule and/or coreceptor. It is differentially expressed by T cells, immature thymocytes, B cells, dendritic cells, macrophages, and endothelial cells. The expression pattern differs according to cell type and activation status. Expression is rapidly induced upon activation of T cells, B cells and dendritic cells, with synthesis by T cells being maintained on Th1 but not Th2 clones. CD150-mediated co-stimulation of TCR-activated T cells enhances the production of IFN-γ by Th1 cells, a response that can be augmented by binding of the TC15-12F12.2 antibody. CD150 is thought to mediate signal transduction by associating with the intracellular protein tyrosine phosphatase, SHP-2. CD150 also has functions in hematopoietic cell development and is a useful marker for detection of multipotent hematopoietic stem cells (in concert with other markers such as CD48 and CD41). CD150 is not expressed on non-multipotent hematopoietic progenitor cells.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD150 (SLAM) Rat Monoclonal Antibody [clone: TC15-12F12.2]
Antigen: CD150 (SLAM)
Clonality: Monoclonal
Clone: TC15-12F12.2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a lambda
Reactivity: Mouse