- Antigen name:TER119
- Clonality:Monoclonal
- Clone:TER-119
- Host:Rat
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Size:100 µg
- Gene ID:104231
- Cat. no.:MSPP-60033
Rat monoclonal IgG2b antibody against mouse TER119, unconjugated.
- Erythroid cells
- Red Blood cells
The TER-119 antibody reacts with murine TER119 (Ly-76), an ~52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs. TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid). The TER-119 antibody is a component of the "lineage cocktail" used to detect, or deplete cells committed to hematopoietic lineages. In adult mice, TER119 is found on ~20 to 25% of bone marrow cells and ~2 to 3% of splenocytes. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ mouse CD4+ T cell isolation kit (Catalog #19852).
Type: Anti-TER119 Rat Monoclonal Antibody [clone: TER-119]
Antigen: TER119
Clonality: Monoclonal
Clone: TER-119
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse