Order Entry
Canada
ContactUsLinkComponent
Anti-CD8a Rat Monoclonal Antibody [clone: 2.43] (APC)
Anti-CD8a Rat Monoclonal Antibody [clone: 2.43] (APC)
Catalog # MSPP-100-1634
undefined
Anti-CD8a Rat Monoclonal Antibody [clone: 2.43] (APC)
Catalog # MSPP-100-1634
Supplier Number:  100-1634

Specifications

  • Antigen name:
    CD8a
  • Clonality:
    Monoclonal
  • Clone:
    2.43
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Size:
    100 μg
  • Gene ID:
    12525
  • Cat. no.:
    MSPP-100-1634

Specifications

About this item

Rat monoclonal antibody against mouse CD8a, APC-conjugated.

  • T cells CD8

This monoclonal antibody (clone CD8a) reacts with mouse cluster of differentiation 8 subunit alpha (CD8a), a 32 - 34 kDa type I transmembrane glycoprotein. CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e. αβ) or a homodimer (αα). Use this recombinant monoclonal antibody in applications requiring detection of CD8a, such as flow cytometry (FC). CD8 is expressed in the αβ form by a majority of thymocytes and a subset of mature peripheral blood T cells (T cytotoxic cells), and in the ββ form by γδ T cells, a subset of intestinal intraepithelial lymphocytes, and dendritic cells. CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding to the major histocompatibility (MHC) Class I molecules presented by an antigen-presenting cell. The CD8a chain binds to the alpha3 domain of MHC class I molecules. It functions to strengthen the association between the TCR and MHC I-antigen complex and to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases, such as lymphocyte-specific protein tyrosine kinase (Lck). This antibody is used as a phenotypic marker for mouse CD8 on cytotoxic T cells, thymocytes, as well as on certain cell types that do not express the TCR, which includes some natural killer cells and lymphoid dendritic cells.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: APC
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse