- Antigen name:IL-4
- Clonality:Monoclonal
- Clone:11B11
- Conjugation:PE (Phycoerythrin)
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:100 μg
- Gene ID:16189
- Cat. no.:MSPP-100-1619
Rat monoclonal antibody against mouse IL-4, PE-conjugated.
- Myeloid cells
- T cells
This monoclonal antibody reacts with mouse interleukin-4 (IL-4), a 14 kDa cytokine that is expressed by Th2-biased CD4+ T cells, mast cells, basophils, and eosinophils. IL-4 is a lymphoid cell growth factor and signals through the IL-4 receptor alpha chain (IL-4Rα) upon binding. This causes the stimulation of activated B and T lymphocytes as well as the differentiation of B cells into plasma cells. Use this recombinant monoclonal antibody in applications requiring detection of IL-4, such as flow cytometry (FC), and the unconjugated antibody in immunocytochemistry (ICC), immunohistochemistry - frozen sections (IHC-F), and FC. IL-4 has the capability to attach and transmit signals through three different cell surface receptors: CD124, CD124 in combination with the common gamma chain (type I complex), or CD124 combined with CD213a1 (type II complex). Signal transducer and activator of transcription 6 (STAT6) has been shown to play an important role in mediating the immune regulatory signal of IL-4. IL-4 has been shown to play a role in normal wound healing and the development of allergic inflammation and asthma.
Type:
Antigen: IL-4
Clonality: Monoclonal
Clone: 11B11
Conjugation: PE
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse