Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: TH-2]
MSPP-60058
- Antigen name:Tyrosine Hydroxylase
- Clonality:Monoclonal
- Clone:TH-2
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat
- Size:200 μl
- Gene ID:7054 (human), 25085 (rat)
- Cat. no.:MSPP-60058
Mouse monoclonal IgG1 antibody against human, rat, cow tyrosine hydroxylase, unconjugated.
- Neural cells Psc derived
The TH-2 antibody clone recognizes an epitope present in the N-terminal region of both rodent (~60 kDa) and human (62 - 68 kDa) tyrosine hydroxylase (TH). TH catalyzes the hydroxylation of L-tyrosine to L-3,4 dihydroxyphenylalanine (L-dopa) in brain and adrenal medulla and can therefore be used to detect dopaminergic neurons. L-dopa is required for the biosynthesis of catecholamines (dopamine, norepinephrine and epinephrine), which function as neurotransmitters and hormones.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Type:
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: TH-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Rat