Order Entry
Canada
ContactUsLinkComponent
Monarch StabiLyse™ DNA/RNA Buffer
Monarch StabiLyse™ DNA/RNA Buffer
Catalog # CA77532-260
CAS Number:  
Monarch StabiLyse™ DNA/RNA Buffer
Catalog # CA77532-260
Supplier Number:  T2111L
CAS Number:  

Specifications

  • Description:
    DNA/RNA buffer
  • Size:
    145 ml
  • Storage temperature:
    25 °C: up to 1 week
    4 °C: up to 4 weeks
    −20 or −80 °C: 4 weeks or more
  • Cat. no.:
    CA77532-260
  • Application:
    DNA and RNA extractions

Specifications

About this item

The Monarch StabiLyse DNA/RNA Buffer is a dual-function reagent for nucleic acid stabilization and sample lysis.

  • Dual-functioning DNA/RNA preservation and sample storage and lysis solution
  • Helps to protect and stabilize RNA and DNA in samples
  • Minimize the need for immediate sample processing
  • Compatible with nucleic acid extraction without need for sample precipitation or reagent removal

This proprietary formulation preserves the integrity of DNA and RNA in samples during storage and it also enables effective sample lysis from a wide variety of eukaryotic and prokaryotic sample types. Samples can be stored in the Monarch StabiLyse DNA/RNA Buffer at the point of harvest for short and long term without the need for immediate processing. The Monarch StabiLyse DNA/RNA Buffer is unique in that it does not require removal of this reagent or sample precipitation. The Monarch StabiLyse DNA/RNA Buffer is ideal for performing sample homogenization and mechanical disruption. The extracted nucleic acids are suitable for a variety of downstream applications including PCR, sequencing, and more.