Specifications
- Concentration:75000 units/ml
- Description:WarmStart RTx reverse transcriptase, glycerol-free
- Cat. no.:CA77532-246
- Units:2000
Specifications
About this item
WarmStart RTx Reverse Transcriptase (Glycerol-free) is formulated without glycerol to support lyophilization, incorporation into microfluidic devices, and enable quick adoption into automation workflows.
- Glycerol-free format to support lyophilization and automation workflows
- Ideal for use in Loop-Mediated Isothermal Amplification (LAMP)
- WarmStart® technology enables room temperature setup and increases consistency and specificity
WarmStart® RTx Reverse Transcriptase is a unique in silico designed RNA-directed DNA polymerase coupled with a reversibly-bound aptamer that inhibits RTx activity below 40 °C. This enzyme can synthesize a complementary DNA strand initiating from a primer using RNA (cDNA synthesis) or single-stranded DNA as a template. RTx is a robust enzyme for RNA detection in amplification reactions and is particularly well suited for use in LAMP (loop-mediated isothermal amplification). The WarmStart® property enables high throughput applications, room temperature setup, and increases the consistency and specificity of amplification reactions. RTx contains intact RNase H activity.