HEPES Buffer Solution (1 M)
Catalog # MSPP-7200
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Description:Biological buffer for cell culture media
- Cat. no.:MSPP-7200
- Volume:100 ml
Specifications
About this item
HEPES Buffer Solution is a biological buffer used in cell culture media.
Selection of suitable nutrient medium is dependent on type of cell, conditions of culture and degree of chemical definition required for the cell culture application.
Species: Human, mouse, non human primate, rat and others.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').