Order Entry
Canada
ContactUsLinkComponent
Human Recombinant FGF-10 (KGF-2)
Human Recombinant FGF-10 (KGF-2)
Catalog # MSPP-780371006
CAS Number:  
Human Recombinant FGF-10 (KGF-2)
Catalog # MSPP-780371006
Supplier Number:  78037.1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/peptide name:
    FGF-10 (KGF-2)
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-780371006

Specifications

About this item

Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family predominantly expressed by mesenchymal fibroblasts during embryonic development (Emoto et al.; Igarashi et al.). It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb (Beer et al.). FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix (Emoto et al.; Igarashi et al.). FGF-10 has been shown to mediate epithelial-mesenchymal interactions, which are essential to lung development (Sekine et al; Ware and Matthay). FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury (Tong et al.; Ware and Matthay), as well as endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells (Takeuchi et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').