Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IL-1 beta, ACF
Human Recombinant IL-1 beta, ACF
Catalog # MSPP-781431006
CAS Number:  
Human Recombinant IL-1 beta, ACF
Catalog # MSPP-781431006
Supplier Number:  78143.1
CAS Number:  

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    100 µg
  • Protein/peptide name:
    IL-1 beta, ACF
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-781431006

Specifications

About this item

Interleukin 1 beta (IL-1β) is synthesized as an inactive precursor protein or pro-IL-1β. This precursor is cleaved intracellularly by Caspase 1 (IL-1β convertase) to form the active form of the protein that is later secreted (Allan et al.). IL-1β binds to IL-1 receptor and activates intracellular signaling via the MAPK or NF-kB pathway. IL-1β is released by monocytes, tissue macrophages, and dendritic cells in response to infection or injury and induces expression of acute-phase proteins. It also promotes the infiltration of inflammatory and immunocompetent cells from the circulation into the extravascular space and affected tissues, by stimulating the expression of adhesion molecules on endothelial cells. IL-1β also affects other immune cells; for example, it co-stimulates T cell functions together with antigen or mitogen. It also stimulates Th17 differentiation and B cell proliferation in an IL-6-dependent manner. This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').