Order Entry
Canada
ContactUsLinkComponent
Human Recombinant G-CSF, ACF
Human Recombinant G-CSF, ACF
Certificates
undefined
Human Recombinant G-CSF, ACF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    G-CSF, ACF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together