Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant Betacellulin
  MSPP-781051006
 :  
undefined
Human Recombinant Betacellulin
  MSPP-781051006
 :  78105.1
 :  

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/peptide name:
    Betacellulin
  • Purity:
    ≥95%
  • Cat. no.:
    MSPP-781051006

 

 

Betacellulin is a member of the epidermal growth factor (EGF) family, and signals through EGF receptor and ERBB4. It activates ERK and AKT pathways, which induces neural stem cell proliferation and prevents spontaneous differentiation in culture. Betacellulin stimulates the expansion of neural stem cells, transit-amplifying cells, and neuroblasts derived from subventricular zone and dentate gyrus (Gómez-Gaviro et al.). It is a potent mitogen for retinal pigment epithelial cells and vascular smooth muscle cells. Betacellulin down-regulates E-cadherin expression in ovarian cancer cell lines via MEK/ERK1/2 and PI3K/AKT signaling pathways, thus increasing cell migration (Zhao et al.). It is a modulator of interferon (IFN) response and enhances anti-viral effects of IFN (Al-Yahya et al.). Betacellulin is expressed in pancreatic α cells, β cells, and duct cells. It induces the proliferation of pancreatic cancer cell lines, inhibits apoptosis, promotes the neogenesis of β cells, and converts non-β cells into insulin-producing cells (Kawaguchi et al.; Miyagawa al.; Saito et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').