Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Size:1000 µg
- Protein/peptide name:PDGF-AA
- Purity:≥95%
- Cat. no.:MSPP-780953
Specifications
About this item
Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark and Heldin). PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin, such as fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). It has been suggested that PDGF-AA is an important autocrine regulator of vascular endothelial growth factor (VEGF) expression in non-small cell lung carcinomas (Shikada et al.). PDGF-AA also mediates proliferation of oligodendrocyte progenitor cells and oligodendrocyte lineage differentiation through the activation of extracellular signal-regulated kinases 1 and 2 (ERK1/2) (Hu et al.). PDGF-AA is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into oligodendrocyte precursor cells (Piao et al.).