About this item
Fibroblast growth factor 10 (FGF-10) is a member of the fibroblast growth factor (FGF) family which is predominantly expressed by mesenchymal fibroblasts during embryonic development (Emoto et al.; Igarashi et al.). It binds with high affinity to fibroblast growth factor receptor 2-IIIb (FGFR2-IIIb), and also has a weaker affinity for FGFR1-IIIb (Beer et al.). FGF-10 and FGF-7 have similar receptor binding properties and target cell specificities but are differentially regulated by components of the extracellular matrix (Emoto et al.; Igarashi et al.). FGF-10 has been shown to mediate epithelial-mesenchymal interactions, which are essential to lung development (Sekine et al.; Ware and Matthay). FGF-10 also has a role in mobilization and proliferation of lung-resident mesenchymal stem cells (MSCs) and protection and repair against acute lung injury (Tong et al.; Ware and Matthay) and endodermal differentiation of human pluripotent stem cells to insulin-producing pancreatic-like cells (Takeuchi et al.). This product is animal component-free.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:FGF-10 (KGF-2), ACF
- Purity:≥95%