Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant IL-10, ACF
 
Human Recombinant IL-10, ACF

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    IL-10, ACF
  • Purity:
    ≥95%

 

Frequently Bought Together