Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IL-1 alpha
Mouse Recombinant IL-1 alpha
Certificates
Mouse Recombinant IL-1 alpha
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-1 alpha
  • Purity:
    ≥95%

Specifications

Frequently Bought Together